Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Amantadine sulfate for synthesis, Sigma-Aldrich®

Supplier: MilliporeSigma
Synonyms: 1-Aminoadamantane sulphate1-Adamantanammonium sulphate1-Adamantylamine sulfate1-Adamantanamine sulfate1-Aminoadamantane sulfate1-Adamantanamine sulphate1-Adamantylamine sulphate1-Adamantanammonium sulfate

M-Clarity™ quality level = MQ200

8.18383.0100 8.18383.0100
EM8.18383.0100CS 1894 USD InStock
CA8.18383.0100 EM8.18383.0100
Amantadine sulfate for synthesis, Sigma-Aldrich®
Amantadine sulphate


Formula: C₂₀H₃₆NO₄S
MW: 386.58 g/mol
MDL Number: MFCD00077251
CAS Number: 31377-23-8
Order Now
Specification Test Results
Solubility (20 °C) insoluble
Melting Point 299 - 301 °C
Molar Mass 400.57 g/mol
pH 4.9 (100 g/l, H2O, 20 °C) (slurry)
Synonyms 1-Aminoadamantane sulfate, Adamantylamine sulfate
Hill Formula C20H36N2O4S
Chemical Formula C20H34N2 * H2SO4

 

ORDER

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty