Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

N,N-Dimethylglycine hydrochloride for synthesis, Sigma-Aldrich®

Supplier: MilliporeSigma

M-Clarity™ quality level = MQ200

8.18448.0010 8.18448.0010
EM8.18448.0010CS 98703 USD InStock
EM8.18448.0010 CA8.18448.0010
N,N-Dimethylglycine hydrochloride for synthesis, Sigma-Aldrich®
N,N-Dimethylglycine hydrochloride


Formula: C₄H₁₀ClNO₂
MDL Number: MFCD00012610
CAS Number: 2491-06-7
Order Now
Specification Test Results
Solubility (20 °C) soluble
Melting Point 190 - 193 °C
Molar Mass 139.58 g/mol
Synonyms N,N-Dimethylaminoacetic acid hydrochloride
Hill Formula C4H10ClNO2
Chemical Formula (CH3)2NCH2COOH * HCl

 

ORDER

DOCUMENTATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty