Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

1-Chlorohexane for synthesis, Sigma-Aldrich®

Supplier: MilliporeSigma
Warning

M-Clarity™ quality level = MQ200

8.18497.0005 8.18497.0250 8.18497.0250 8.18497.0005
EM8.18497.0005CS 2013 USD InStock
CA8.18497.0250 EM8.18497.0005 CA8.18497.0005 EM8.18497.0250
1-Chlorohexane for synthesis, Sigma-Aldrich®
1-Chlorohexane


Formula: C₆H₁₃Cl
MW: 120.62 g/mol
MDL Number: MFCD00001018
CAS Number: 544-10-5
UN: 1993
ADR: 3,II
Order Now
Specification Test Results
Solubility 0.26 g/l (20 °C)
Melting Point -94 °C
Molar Mass 120.62 g/mol
Boiling Point 132 - 136 °C (1013 hPa)
Vapor Pressure 53 hPa (55 °C)
Flash Point 27 °C
Refractive Index 1.4194 (20 °C, 589 nm)
Explosion Limit 1.0 - 9.6 %(V)
Density 0.88 g/cm3 (20 °C)
Ignition Point 225 °C

 

ORDER

DOCUMENTATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty