Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

1,2,3,4,5-Pentamethylcyclopentadiene for synthesis, Sigma-Aldrich®

Supplier: MilliporeSigma
Warning
Synonyms: 1,2,3,4,5-Pentamethylcyclopenta-1,3-diene1,2,3,4,5-Pentamethyl-1,3-cyclopentadienePentamethylcyclopentadiene

M-Clarity™ quality level = MQ200

8.18534.0005 8.18534.0005 8.18534.0001 8.18534.0001
EM8.18534.0005CS 199535 USD InStock
EM8.18534.0005 CA8.18534.0001 CA8.18534.0005 EM8.18534.0001
1,2,3,4,5-Pentamethylcyclopentadiene for synthesis, Sigma-Aldrich®
1,2,3,4,5-Pentamethylcyclopentadiene


Formula: C₁₀H₁₆
MW: 136.24 g/mol
MDL Number: MFCD00001354
CAS Number: 4045-44-7
UN: 1993
ADR: 3,III
Order Now
Specification Test Results
Molar Mass 136.23 g/mol
Boiling Point 58 °C (17 hPa)
Flash Point 44 °C
Refractive Index 1.474 (20 °C, 589 nm)
Density 0.84 g/cm3 (20 °C)

 

ORDER

DOCUMENTATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty