Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

4'-Nitrocinnamic acid for synthesis, Sigma-Aldrich®

Supplier: MilliporeSigma
Synonyms: trans-4-Nitrocinnamic acidtrans-3-(4-Nitrophenyl)propenoic acid

M-Clarity™ quality level = MQ200

8.18563.0025 8.18563.0025
EM8.18563.0025CS 11361 USD InStock
EM8.18563.0025 CA8.18563.0025
4'-Nitrocinnamic acid for synthesis, Sigma-Aldrich®
4'-Nitrocinnamic acid


Formula: C₉H₇NO₄
MW: 193.16 g/mol
MDL Number: MFCD00007381
CAS Number: 882-06-4
Order Now
Specification Test Results
Solubility (20 °C) insoluble
Melting Point 288 - 291 °C
Molar Mass 193.15 g/mol
Synonyms trans-3-(4-Nitrophenyl)propenoic acid
Hill Formula C9H7NO4
Chemical Formula 4-(NO2)C6H4CH=CHCOOH

 

ORDER

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty