Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Nicotinic acid (Niacin) for synthesis, Sigma-Aldrich®

Supplier: MilliporeSigma
Warning
Synonyms: Pyridine-3-carboxylic acid3-Pyridinecarboxylic acidVitamin B33-Picolinic acidNicotinic acidNiacin

M-Clarity™ quality level = MQ200

8.18714.1000 8.18714.1000 8.18714.0100 8.18714.0100
EM8.18714.1000CS 1057 USD InStock
EM8.18714.1000 EM8.18714.0100 CA8.18714.1000 CA8.18714.0100
Nicotinic acid (Niacin) for synthesis, Sigma-Aldrich®
Nicotinic acid (Niacin)


Formula: C₆H₅NO₂
MW: 123.11 g/mol
MDL Number: MFCD00006391
CAS Number: 59-67-6
Order Now
Specification Test Results
Assay (acidimetric) ≥ 99.0 %
Identity (IR) passes test
Melting range (lower value) ≥ 234 °C
Melting range (upper value) ≤ 238 °C

 

ORDER

DOCUMENTATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty