Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

2,3-Butanediol for synthesis, Sigma-Aldrich®

Supplier: MilliporeSigma
Synonyms: 2,3-Dihydroxy butane2,3-Butylene glykol(±)-2,3-Butanediol

M-Clarity™ quality level = MQ200

8.18801.1000 8.18801.0250 8.18801.9010 8.18801.9010 8.18801.0005 8.18801.0005 8.18801.0250 8.18801.1000
EM8.18801.0250CS 1283 USD InStock
CA8.18801.0005 EM8.18801.0250 EM8.18801.9010 CA8.18801.0250 CA8.18801.1000 EM8.18801.0005 EM8.18801.1000 CA8.18801.9010
2,3-Butanediol for synthesis, Sigma-Aldrich®
2,3-Butanediol
Mixture of meso- D- and L-form


Formula: C₄H₁₀O₂
MW: 90.12 g/mol
Boiling Pt: 184 °C (1013 hPa)
Melting Pt: 18…20 °C
Density: 1 g/cm³ (20 °C)
Storage Temperature: Ambient
MDL Number: MFCD00004523
CAS Number: 513-85-9
EINECS: 208-173-6
Order Now
Specification Test Results
Solubility 1000 g/l (20°C)
Melting Point 18 - 20 °C
Boiling Point 180 - 182 °C (1013 hPa)
Vapor Pressure  - 1 hPa (20 °C)
Flash Point 96 °C DIN 51758
Refractive Index 1.4348 (20 °C, 589 nm)
Explosion Limit 3.1 - 11.4 %(V)
Density 1.00 g/cm3 (20 °C)
pH 7 (H2O, 20 °C) Aqueous solution
Ignition Point 395°C DIN 51794

 

ORDER

DOCUMENTATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty