Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Dibutyl disulfide for synthesis, Sigma-Aldrich®

Supplier: MilliporeSigma
Warning
Synonyms: n-Butyl disulphiden-Butyl disulfide

M-Clarity™ quality level = MQ200

8.20242.0025 8.20242.0025 8.20242.0100 8.20242.0100
EM8.20242.0100CS 2460 USD InStock
CA8.20242.0100 CA8.20242.0025 EM8.20242.0100 EM8.20242.0025
Dibutyl disulfide for synthesis, Sigma-Aldrich®
Dibutyl disulphide


Formula: C₈H₁₈S₂
MW: 178.36 g/mol
MDL Number: MFCD00009467
CAS Number: 629-45-8
Order Now
Specification Test Results
Solubility (20 °C) insoluble
Melting Point -94 °C
Molar Mass 178.35 g/mol
Boiling Point 229 - 233 °C
Flash Point 93 °C
Refractive Index 1.492 (20 °C, 589 nm)
Explosion Limit 0.8 %(V)
Density 0.94 g/cm3 (20 °C)
Synonyms Butyl disulfide
Hill Formula C8H18S2
Chemical Formula CH3(CH2)3SS(CH2)3CH3

 

ORDER

DOCUMENTATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty