Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Dibutyltin diacetate for synthesis, Sigma-Aldrich®

Supplier: MilliporeSigma
Danger
Synonyms: Di-n-butyltin diacetate

M-Clarity™ quality level = MQ200

8.20386.0100 8.20386.9010 8.20386.9010 8.20386.0100
EM8.20386.0100CS 2005 USD InStock
EM8.20386.0100 CA8.20386.9010 EM8.20386.9010 CA8.20386.0100
Dibutyltin diacetate for synthesis, Sigma-Aldrich®
Dibutyltin diacetate


Formula: C₁₂H₂₄O₄Sn
MW: 351.03 g/mol
MDL Number: MFCD00008697
CAS Number: 1067-33-0
UN: 2788
ADR: 6.1,II
Order Now
Specification Test Results
Solubility 0.006 g/l (20 °C)
Melting Point 12 °C
Molar Mass 351 g/mol
Boiling Point 142 - 145 °C (13 hPa)
Vapor Pressure 1.7 hPa (25 °C)
Flash Point 143 °C
Density 1.30 g/cm3 (20 °C)
Ignition Point 200 °C

 

ORDER

DOCUMENTATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty