Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

meso-Hydrobenzoin for synthesis, Sigma-Aldrich®

Supplier: MilliporeSigma
Synonyms: rel-(1R,2S)-1,2-Diphenylethane-1,2-diolmeso-1,2-Diphenyl-1,2-ethanediolmeso-1,2-Diphenylethylene glycol

M-Clarity™ quality level = MQ200

8.20526.0001 8.20526.0001
EM8.20526.0001CS 106008 USD InStock
CA8.20526.0001 EM8.20526.0001
meso-Hydrobenzoin for synthesis, Sigma-Aldrich®
meso-Hydrobenzoin


Formula: C₁₄H₁₄O₂
MW: 214.26 g/mol
MDL Number: MFCD00004499
CAS Number: 579-43-1
Order Now
Specification Test Results
Solubility (20 °C) slightly soluble
Melting Point 128 - 131 °C
Molar Mass 214.26 g/mol
Synonyms meso-Hydrobenzoin, meso-1,2-Diphenylethylene glycol
Hill Formula C14H14O2
Chemical Formula HOCH(C6H5)CH(C6H5)OH

 

ORDER

DOCUMENTATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty