Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

N,N'-Diphenyl-p-phenylenediamine for synthesis, Sigma-Aldrich®

Supplier: MilliporeSigma
Warning
Synonyms: N,N'-Diphenyl-1,4-phenylenediamine

M-Clarity™ quality level = MQ200

8.20536.0250 8.20536.9010 8.20536.9010 8.20536.0250
EM8.20536.0250CS 3199 USD InStock
CA8.20536.0250 CA8.20536.9010 EM8.20536.0250 EM8.20536.9010
N,N'-Diphenyl-p-phenylenediamine for synthesis, Sigma-Aldrich®
N,N'-Diphenyl-p-phenylenediamine


Formula: C₁₈H₁₆N₂
MW: 260.34 g/mol
MDL Number: MFCD00003015
CAS Number: 74-31-7
Order Now
Specification Test Results
Solubility (20 °C) practically insoluble
Melting Point 135 °C
Molar Mass 260.33 g/mol
Bulk Density 560 kg/m3
Boiling Point 220 - 225 °C (1.33 hPa)
Density 1.3 g/cm3 (20 °C)

 

ORDER

DOCUMENTATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty