Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Isatin for synthesis, Sigma-Aldrich®

Supplier: MilliporeSigma
Synonyms: Indoline-2,3-dione2,3-Indolinedione1H-Indole-2,3-dione

M-Clarity™ quality level = MQ200

8.20709.0250 8.20709.0250 8.20709.0005 8.20709.0005
EM8.20709.0250CS 3140 USD InStock
CA8.20709.0005 EM8.20709.0250 CA8.20709.0250 EM8.20709.0005
Isatin for synthesis, Sigma-Aldrich®
Isatin


Formula: C₈H₅NO₂
MW: 147.13 g/mol
MDL Number: MFCD00005718
CAS Number: 91-56-5
Order Now
Specification Test Results
Solubility 2 g/l (20 °C)
Melting Point 199 - 204 °C
Molar Mass 147.13 g/mol
Bulk Density 600 kg/m3
Flash Point 200 °C
pH 7 (1.9 g/l, H2O, 20 °C)
Ignition Point 220 °C
Hill Formula C8H5NO2

 

ORDER

DOCUMENTATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty