Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Sodium phenoxide trihydrate for synthesis, Sigma-Aldrich®

Supplier: MilliporeSigma
Danger
Synonyms: Sodium phenolate trihydrate

M-Clarity™ quality level = MQ200

8.20873.0500 8.20873.0100 8.20873.0100 8.20873.0500
EM8.20873.0100CS 499 USD InStock
CA8.20873.0500 EM8.20873.0100 EM8.20873.0500 CA8.20873.0100
Sodium phenoxide trihydrate for synthesis, Sigma-Aldrich®
Sodium phenoxide trihydrate


Formula: C₆H₅NaO·3H₂O
MDL Number: MFCD00149636
CAS Number: 156150-40-2
UN: 2905
ADR: 8,III
Order Now
Specification Test Results
Solubility (20 °C) soluble
Melting Point 56 - 61 °C
Molar Mass 170.13 g/mol

 

ORDER

DOCUMENTATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty