Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Ethyl pelargonate for synthesis, Sigma-Aldrich®

Supplier: MilliporeSigma
Synonyms: Ethyl nonanoatePelargonic acid ethyl esterNonanoic acid ethyl ester

M-Clarity™ quality level = MQ200

8.20916.0250 8.20916.0250
EM8.20916.0250CS 1310 USD InStock
EM8.20916.0250 CA8.20916.0250
Ethyl pelargonate for synthesis, Sigma-Aldrich®
Ethyl pelargonate


Formula: C₁₁H₂₂O₂
MW: 186.29 g/mol
MDL Number: MFCD00009570
CAS Number: 123-29-5
Order Now
Specification Test Results
Solubility (20 °C) insoluble
Melting Point -44 °C
Molar Mass 186.29 g/mol
Boiling Point 220 - 222 °C
Flash Point 93 °C
Density 0.86 g/cm3 (20 °C)
Synonyms Nonanoic acid ethyl ester, Ethyl pelargonate, Pelargonic acid ethyl ester
Hill Formula C11H22O2
Chemical Formula CH3(CH2)7COOC2H5

 

ORDER

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty