Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Phosphorus pentasulfide for synthesis, Sigma-Aldrich®

Supplier: MilliporeSigma
Danger
Synonyms: di-Phosphorus pentasulphidedi-Phosphorus pentasulfide

M-Clarity™ quality level = MQ200

8.21024.1000 8.21024.2500 8.21024.0100 8.21024.2500 8.21024.0100 8.21024.1000
EM8.21024.1000CS 1669 USD InStock
CA8.21024.0100 CA8.21024.2500 EM8.21024.1000 EM8.21024.0100 EM8.21024.2500 CA8.21024.1000
Phosphorus pentasulfide for synthesis, Sigma-Aldrich®
Phosphorus pentasulphide


Formula: P₂S₅
MW: 222.28 g/mol
MDL Number: MFCD00011441
CAS Number: 1314-80-3
UN: 1340
ADR: 4.3,II
Order Now
Specification Test Results
Solubility (20 °C) (decomposition)
Melting Point 284 - 287 °C
Molar Mass 222.28 g/mol
Bulk Density 900 - 1100 kg/m3
Boiling Point 513 - 515 °C (1013 hPa)
Density 2.08 g/cm3 (20 °C)
pH 1 (10 g/l, H2O, 20 °C)
Ignition Point 142 °C
Hazard Class 4.3
Hazard Class Description Dangerous When Wet Material

 

ORDER

DOCUMENTATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty