Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Trimethyl borate for synthesis, Sigma-Aldrich®

Supplier: MilliporeSigma
Danger

M-Clarity™ quality level = MQ200

8.21180.1000 8.21180.1000 8.21180.0250 8.21180.0250
EM8.21180.1000CS 1219 USD InStock
CA8.21180.0250 CA8.21180.1000 EM8.21180.1000 EM8.21180.0250
Trimethyl borate for synthesis, Sigma-Aldrich®
Trimethyl borate


Formula: C₃H₉BO₃
MW: 103.91 g/mol
Boiling Pt: 67 °C (1013 hPa)
Melting Pt: –5 °C
Density: 0.915 g/cm³ (20 °C)
Flash Pt: –11 °C
Storage Temperature: Refrigerator
MDL Number: MFCD00008346
CAS Number: 121-43-7
EINECS: 204-468-9
UN: 2416
ADR: 3,II
Merck Index: 12,09842
Order Now
Specification Test Results
Solubility (20 °C) (decomposition)
Melting Point -34 - -29 °C
Molar Mass 103.91 g/mol
Boiling Point 67 - 69 °C (1013 hPa)
Vapor Pressure 182 hPa (25 °C)
Flash Point -8 °C
Refractive Index 1.3568 (20 °C)
Explosion Limit 0.932 g/cm3 (20 °C)
Density (H2O) no data available
Ignition Point 305 °C
Hazard Class 3
Hazard Class Description Flammable Liquid

 

ORDER

DOCUMENTATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty