Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Trimethyl phosphate for synthesis, Sigma-Aldrich®

Supplier: MilliporeSigma
Danger
Synonyms: Trimethylphosphate

M-Clarity™ quality level = MQ200

8.21184.0500 8.21184.0100 8.21184.0500 8.21184.2500 8.21184.0100 8.21184.2500
EM8.21184.0500CS 502 USD InStock
CA8.21184.0100 EM8.21184.0500 EM8.21184.0100 EM8.21184.2500 CA8.21184.0500 CA8.21184.2500
Trimethyl phosphate for synthesis, Sigma-Aldrich®
Trimethyl phosphate


Formula: C₃H₉O₄P
MW: 140.08 g/mol
Boiling Pt: 197.2 °C (1013 hPa)
Melting Pt: –46 °C
Density: 1.21 g/cm³ (20 °C)
Flash Pt: >150 °C
Storage Temperature: Ambient
MDL Number: MFCD00008348
CAS Number: 512-56-1
EINECS: 208-144-8
Order Now
Specification Test Results
Solubility 1000 g/l (25 °C)
Melting Point -46 °C
Molar Mass 140.08 g/mol
Boiling Point 197 °C (1013 hPa)
Vapor Pressure 1.133 hPa (25 °C)
Flash Point 150 °C
Refractive Index 1.3967 (20 °C)
Density 1.21 g/cm3 (20 °C)

 

ORDER

DOCUMENTATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty