Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

2,5-Di-tert-butylhydroquinone for synthesis, Sigma-Aldrich®

Supplier: MilliporeSigma
Danger
Synonyms: BHQ

M-Clarity™ quality level = MQ200

8.21762.0250 8.21762.1000 8.21762.1000 8.21762.0250
EM8.21762.0250CS 1341 USD InStock
CA8.21762.0250 CA8.21762.1000 EM8.21762.0250 EM8.21762.1000
2,5-Di-tert-butylhydroquinone for synthesis, Sigma-Aldrich®
2,5-Di-tert-butylhydroquinone


Formula: C₁₄H₂₂O₂
MW: 222.33 g/mol
Boiling Pt: 334 °C (1013 hPa)
Melting Pt: 212 °C
Density: 1.078 g/cm³ (20 °C)
Storage Temperature: Ambient
MDL Number: MFCD00008825
CAS Number: 88-58-4
EINECS: 201-841-8
UN: 2811
ADR: 6.1,III
Order Now
Specification Test Results
Solubility (20 °C) insoluble
Melting Point 214 - 217 °C
Molar Mass 222.32 g/mol
Ignition Point 420 °C

 

ORDER

DOCUMENTATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty