Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Propyl benzoate for synthesis, Sigma-Aldrich®

Supplier: MilliporeSigma
Warning
Synonyms: Benzoic acid propyl ester

M-Clarity™ quality level = MQ200

8.21958.0250 8.21958.0005 8.21958.0005 8.21958.0250
EM8.21958.0005CS 2565 USD InStock
CA8.21958.0250 EM8.21958.0005 CA8.21958.0005 EM8.21958.0250
Propyl benzoate for synthesis, Sigma-Aldrich®
Propyl benzoate


Formula: C₁₀H₁₂O₂
MW: 164.2 g/mol
Boiling Pt: 231 °C (1013 hPa)
Melting Pt: –52 °C
Density: 1.02 g/cm³ (20 °C)
Flash Pt: >100 °C
Storage Temperature: Ambient
MDL Number: MFCD00009370
CAS Number: 2315-68-6
EINECS: 219-020-8
Order Now
Specification Test Results
Solubility 0.35 g/l (25 °C)
Melting Point -52 °C
Molar Mass 164.2 g/mol
Boiling Point 231 °C (1013 hPa)
Vapor Pressure 0.181 hPa (25 °C)
Flash Point 100 °C
Refractive Index 1.4981 (25 °C, 589 nm)
Density 1.02 g/cm3 (20 °C)

 

ORDER

DOCUMENTATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty