Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

2,6-Di-tert-butyl-p-cresol for synthesis, Sigma-Aldrich®

Supplier: MilliporeSigma
Warning
Synonyms: 2,6-Di-tert-butyl-4-methylphenolButylhydroxytolueneButylated hydroxytolueneBHT

M-Clarity™ quality level = MQ200

8.22021.9025 8.22021.0100 8.22021.2500 8.22021.0500 8.22021.9025 8.22021.0100 8.22021.0500 8.22021.2500
EM8.22021.0500CS 1619 USD InStock
CA8.22021.2500 CA8.22021.0100 EM8.22021.0500 CA8.22021.9025 EM8.22021.2500 EM8.22021.0100 EM8.22021.9025 CA8.22021.0500
2,6-Di-tert-butyl-p-cresol for synthesis, Sigma-Aldrich®
2,6-Di-tert-butyl-p-cresol


Formula: C₁₅H₂₄O
MW: 220.35 g/mol
Boiling Pt: 265 °C (1013 hPa)
Melting Pt: 69…70 °C
Density: 1.05 g/cm³ (20 °C)
Flash Pt: 127 °C
Storage Temperature: Ambient
MDL Number: MFCD00011644
CAS Number: 128-37-0
EINECS: 204-881-4
UN: 3077
ADR: 9,III
Merck Index: 13,01547
Order Now
Specification Test Results
Solubility  - 0.001 g/l (25 °C)
Melting Point 69 - 70 °C
Molar Mass 220.35 g/mol
Bulk Density 450 kg/m3
Boiling Point 265 °C (1013 hPa)
Vapor Pressure 0.02 hPa (20 °C)
Flash Point 127 °C
Density 1.05 g/cm3 (20 °C)
pH (H2O) not applicable
Ignition Point 345 °C

 

ORDER

DOCUMENTATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty