Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

4-(Dimethylamino)cinnamaldehyde for synthesis, Sigma-Aldrich®

Supplier: MilliporeSigma
Warning
Synonyms: p-Dimethylamino-Cinnamaldehyde

M-Clarity™ quality level = MQ200

8.22034.0500 8.22034.0010 8.22034.0010 8.22034.0002 8.22034.0002 8.22034.0500
EM8.22034.0010CS 13299 USD InStock
EM8.22034.0010 EM8.22034.0002 CA8.22034.0002 CA8.22034.0010 EM8.22034.0500 CA8.22034.0500
4-(Dimethylamino)cinnamaldehyde for synthesis, Sigma-Aldrich®
4-(Dimethylamino)cinnamaldehyde


Formula: C₁₁H₁₃NO
MW: 175.23 g/mol
MDL Number: MFCD00007002
CAS Number: 6203-18-5
EINECS: 228-267-0
Order Now
Specification Test Results
Solubility (20 °C) insoluble
Melting Point 135 - 138 °C
Molar Mass 175.22 g/mol

 

ORDER

DOCUMENTATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty