Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

1-Dodecene for synthesis, Sigma-Aldrich®

Supplier: MilliporeSigma
Danger
Synonyms: Dodec-1-ene

M-Clarity™ quality level = MQ200

8.22048.0500 8.22048.0500 8.22048.0100 8.22048.0100
EM8.22048.0500CS 759 USD InStock
CA8.22048.0500 EM8.22048.0500 CA8.22048.0100 EM8.22048.0100
1-Dodecene for synthesis, Sigma-Aldrich®
1-Dodecene


Formula: C₁₂H₂₄
MW: 168.32 g/mol
Boiling Pt: 213 °C (1013 hPa)
Melting Pt: –35 °C
Density: 0.7582 g/cm³ (20 °C)
Flash Pt: 77 °C
Storage Temperature: Ambient
MDL Number: MFCD00008961
CAS Number: 112-41-4
EINECS: 203-968-4
Order Now
Specification Test Results
Solubility (20 °C) insoluble
Melting Point -35 °C
Molar Mass 168.32 g/mol
Boiling Point 213 - 216 °C (1013 hPa)
Vapor Pressure 0.23 hPa (20 °C)
Flash Point 77 °C
Refractive Index 1.4294 (20 °C, 589 nm)
Explosion Limit 0.6 - 5.4 %(V)
Density 0.76 g/cm3 (20 °C)
Ignition Point 226 °C
Viscosity kinematic 1.71 mm2/s (20 °C)

 

ORDER

DOCUMENTATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty