Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Scheduled maintenance of the site is planned from June 8th at 7:00 PM to June 9th at 5:00 PM ET. During this time, contract pricing and registration functionality will be unavailable. We appreciate your patience.

Benzoic acid for synthesis, Sigma-Aldrich®

Supplier: MilliporeSigma
Danger

M-Clarity™ quality level = MQ200

8.22257.0100 8.22257.0100 8.22257.5000 8.22257.1000 8.22257.5000 8.22257.1000
EM8.22257.0100CS 2316 USD InStock
EM8.22257.0100 CA8.22257.5000 CA8.22257.1000 CA8.22257.0100 EM8.22257.5000 EM8.22257.1000
Benzoic acid for synthesis, Sigma-Aldrich®
Benzoic acid


Formula: C₇H₆O₂
MW: 122.12 g/mol
Boiling Pt: 249 °C (1013 hPa)
Melting Pt: 121…123 °C
Density: 1.316 g/cm³ (20 °C)
Flash Pt: 121 °C (closed cup)
Storage Temperature: Ambient
MDL Number: MFCD00002398
CAS Number: 65-85-0
EINECS: 200-618-2
REACH: 01-2119455536-33
Merck Index: 13,01092
https://us-legacy.vwr.com/stibo/web/std.lang.all/81/11/748111.jpg
Order Now
Specification Test Results
Assay (acidimetric) ≥ 99.0 %
Identity (IR) passes test
Melting range (lower value) ≥ 122 °C
Melting range (upper value) ≤ 125 °C

 

ORDER

DOCUMENTATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty