Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Phthalic acid for synthesis, Sigma-Aldrich®

Supplier: MilliporeSigma
Warning
Synonyms: Pathalic acid

M-Clarity™ quality level = MQ200

8.22298.1000 8.22298.0100 8.22298.1000 8.22298.0100
EM8.22298.0100CS 6212 USD InStock
CA8.22298.0100 CA8.22298.1000 EM8.22298.0100 EM8.22298.1000
Phthalic acid for synthesis, Sigma-Aldrich®
Phthalic acid


Formula: C₈H₆O₄
MW: 166.13 g/mol
Boiling Pt: 315…317 °C (1013 hPa)
Melting Pt: 79 °C
Density: 1.576 g/cm³ (20 °C)
Storage Temperature: Ambient
MDL Number: MFCD00002467
CAS Number: 88-99-3
EINECS: 201-873-2
Merck Index: 12,07527
Order Now
Specification Test Results
Assay (acidimetric) ≥ 99.0 %
Identity (IR) passes test

 

ORDER

DOCUMENTATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty