Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Cyclohexanol for synthesis, Sigma-Aldrich®

Supplier: MilliporeSigma
Warning
Synonyms: 1-Hydroxycyclohexane

M-Clarity™ quality level = MQ200

8.22328.9025 8.22328.0100 8.22328.9025 8.22328.0100 8.22328.1000 8.22328.1000 8.22328.2500 8.22328.2500
EM8.22328.0100CS 642 USD InStock
CA8.22328.2500 CA8.22328.9025 CA8.22328.1000 CA8.22328.0100 EM8.22328.0100 EM8.22328.2500 EM8.22328.9025 EM8.22328.1000
Cyclohexanol for synthesis, Sigma-Aldrich®
Cyclohexanol


Formula: C₆H₁₂O
MW: 100.16 g/mol
Boiling Pt: 161 °C (1013 hPa)
Melting Pt: 25 °C
Density: 0.947 g/cm³ (20 °C)
Flash Pt: 59 °C
Storage Temperature: Ambient
MDL Number: MFCD00003855
CAS Number: 108-93-0
EINECS: 203-630-6
Merck Index: 13,02754
https://us-legacy.vwr.com/stibo/web/std.lang.all/21/52/772152.jpg
Order Now
Specification Test Results
Solubility 40 g/l (20 °C)
Melting Point 25 °C
Molar Mass 100.16 g/mol
Boiling Point 161 °C (1013 hPa)
Vapor Pressure 0.6 hPa (20 °C)
Refractive Index 1.4656 (20 °C, 589 nm)
Explosion Limit 2 - 11.2 %(V)
Density 0.94 g/cm3 (25 °C)
pH 6.5 (40 g/l, H2O, 20 °C)
Ignition Point 300 °C
Saturation concentration 5 g/m3 (20 °C) Air

 

ORDER

DOCUMENTATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty