Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

m-Xylene for synthesis, Sigma-Aldrich®

Supplier: MilliporeSigma
Warning
Synonyms: 1,3-Dimethylbenzene

M-Clarity™ quality level = MQ200

8.22337.2500 8.22337.1000 8.22337.1000 8.22337.2500
EM8.22337.2500CS 976 USD InStock
CA8.22337.2500 EM8.22337.2500 CA8.22337.1000 EM8.22337.1000
m-Xylene for synthesis, Sigma-Aldrich®
m-Xylene


Formula: C₈H₁₀
MW: 106.17 g/mol
Boiling Pt: 138 °C (1013 hPa)
Melting Pt: –48 °C
Density: 0.864 g/cm³ (20 °C)
Flash Pt: 25 °C
Storage Temperature: Ambient
MDL Number: MFCD00008536
CAS Number: 108-38-3
EINECS: 203-576-3
UN: 1307
ADR: 3,III
Merck Index: 12,10214
Order Now
Specification Test Results
Solubility 0.2 g/l (20 °C)
Melting Point -48 °C
Molar Mass 106.16 g/mol
Boiling Point 139 °C (1013 hPa)
Vapor Pressure 8 hPa (20 °C)
Flash Point 25 °C
Explosion Limit 1.1 - 7 %(V)
Density 0.86 g/cm3 (20 °C)
Ignition Point 525 °C
Saturation concentration 35 g/m3 (20 °C) Air
Evaporation number 13.5
Hazard Class 3
Hazard Class Description Flammable Liquid

 

ORDER

DOCUMENTATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty