Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

2,4-Dichlorobenzonitrile for synthesis, Sigma-Aldrich®

Supplier: MilliporeSigma
Warning

M-Clarity™ quality level = MQ200

8.40037.0100 8.40037.0100
EM8.40037.0100CS 1108 USD InStock
EM8.40037.0100 CA8.40037.0100
2,4-Dichlorobenzonitrile for synthesis, Sigma-Aldrich®
2,4-Dichlorobenzonitrile


Formula: C₇H₃Cl₂N
MW: 172.01 g/mol
Boiling Pt: 261 °C (1013 hPa)
Melting Pt: 52…53 °C
Storage Temperature: Ambient
MDL Number: MFCD00016373
CAS Number: 6574-98-7
EINECS: 229-493-2
ADR: 6.1,III
Order Now
Specification Test Results
Solubility (20 °C) insoluble
Melting Point 59 - 62 °C
Molar Mass 172.01 g/mol

 

ORDER

DOCUMENTATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty