Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

2-Ethoxyethyl methacrylate stabilized for synthesis, Sigma-Aldrich®

Supplier: MilliporeSigma
Warning
Synonyms: Methacrylic acid 2-Ethoxyethyl esterEthylene glycol monoethyl ether Methacrylate

M-Clarity™ quality level = MQ200

8.40085.0025 8.40085.0025
EM8.40085.0025CS 4824 USD InStock
EM8.40085.0025 CA8.40085.0025
2-Ethoxyethyl methacrylate stabilized for synthesis, Sigma-Aldrich®
2-Ethoxyethyl methacrylate


Formula: C₈H₁₄O₃
MW: 158.1950 g/mol
MDL Number: MFCD00009249
CAS Number: 2370-63-0
EINECS: 219-135-3
Order Now
Specification Test Results
Solubility 13 g/l (20 °C)
Molar Mass 158.19 g/mol
Boiling Point 205 °C
Vapor Pressure 1.5 hPa (20 °C)
Flash Point 74 °C
Density 0.96 g/cm3 (20 °C)
Ignition Point 209 °C

 

ORDER

DOCUMENTATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty