Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

N-Methyl-N-(trimethylsilyl)trifluoroacetamide for synthesis, Sigma-Aldrich®

Supplier: MilliporeSigma
Warning
Synonyms: MSTFAN-Methyl-N-(trimethylsilyl)-2,2,2-trifluoroacetamide

M-Clarity™ quality level = MQ200

8.41102.0025 8.41102.0005 8.41102.0025 8.41102.0005
EM8.41102.0005CS 7261 USD InStock
CA8.41102.0025 EM8.41102.0005 EM8.41102.0025 CA8.41102.0005
N-Methyl-N-(trimethylsilyl)trifluoroacetamide for synthesis, Sigma-Aldrich®
N-Methyl-N-(trimethylsilyl)trifluoroacetamide


Formula: C₆H₁₂F₃NOSi
MW: 199.25 g/mol
Boiling Pt: 130 °C (1013 hPa)
Density: 1.07 g/cm³ (20 °C)
MDL Number: MFCD00000411
CAS Number: 24589-78-4
EINECS: 246-331-6
UN: 1993
ADR: 3,II
Order Now
Specification Test Results
Molar Mass 199.25 g/mol
Boiling Point 130 - 132 °C (1013 hPa)
Flash Point 25 °C
Density 1.085 g/cm3

 

ORDER

DOCUMENTATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty