Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

DL-Norvaline ≥98% for synthesis, Sigma-Aldrich®

Supplier: MilliporeSigma
Synonyms: 2-Aminopentanoic acidH-DL-Nva-OH(±)-Norvaline

M-Clarity™ quality level = MQ200

8.41562.0025 8.41562.0025
EM8.41562.0025CS 10378 USD InStock
EM8.41562.0025 CA8.41562.0025
DL-Norvaline ≥98% for synthesis, Sigma-Aldrich®
DL-Norvaline


Formula: C₅H₁₁NO₂
MW: 117.15 g/mol
Boiling Pt: 223 °C (1013 hPa)
Melting Pt: > 300 °C
Storage Temperature: Ambient
MDL Number: MFCD00064420
CAS Number: 760-78-1
EINECS: 212-082-7
Merck Index: 12,06813
Order Now
Specification Test Results
Assay (HClO₄) ≥ 98.0 %
Identity (IR) passes test

 

ORDER

DOCUMENTATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty