Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

2,5-Dihydroxybenzoic acid for synthesis, Sigma-Aldrich®

Supplier: MilliporeSigma
Synonyms: 2,5-DHBA2,5-Dihydroxybenzoic AcidGentisic acidHydroquinonecarboxylic acidDHB

M-Clarity™ quality level = MQ200

8.41745.0050 8.41745.0250 8.41745.0250 8.41745.0050
EM8.41745.0050CS 13930 USD InStock
EM8.41745.0050 EM8.41745.0250 CA8.41745.0250 CA8.41745.0050
2,5-Dihydroxybenzoic acid for synthesis, Sigma-Aldrich®
2,5-Dihydroxybenzoic acid


Formula: C₇H₆O₄
MW: 154.12 g/mol
Boiling Pt: 339 °C (1013 hPa)
Melting Pt: 202…206 °C
Density: 1.55 g/cm³ (20 °C)
Storage Temperature: Ambient
MDL Number: MFCD00002460
CAS Number: 490-79-9
EINECS: 207-718-5
Merck Index: 13,04409
Order Now
Specification Test Results
Melting Point 202 - 206 °C
Molar Mass 154.12 g/mol

 

ORDER

DOCUMENTATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty