Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

1-Bromo-3-fluoro-4-(trifluoromethoxy)benzene for synthesis, Sigma-Aldrich®

Supplier: MilliporeSigma
Warning
Synonyms: 4-Bromo-2-fluoro-1-(trifluoromethoxy)benzene

M-Clarity™ quality level = MQ200

8.43874.0050 8.43874.0010 8.43874.0050 8.43874.0010
EM8.43874.0010CS 328626 USD InStock
EM8.43874.0010 CA8.43874.0050 EM8.43874.0050 CA8.43874.0010
1-Bromo-3-fluoro-4-(trifluoromethoxy)benzene for synthesis, Sigma-Aldrich®
1-Bromo-3-fluoro-4-(trifluoromethoxy)benzene


Formula: C₇H₃BrF₄O
MW: 259.00 g/mol
MDL Number: MFCD01861132
CAS Number: 105529-58-6
UN: 3082
ADR: 9,III
Order Now
Specification Test Results
Solubility 0.0288 g/l (20 °C)
Molar Mass 259 g/mol
Boiling Point 171 °C (994.5 hPa)
Vapor Pressure 4 hPa (25 °C)
Flash Point 69 °C
Density 1.63 g/cm3 (20 °C)
pH (H2O) neutral
Ignition Point 630 °C
Hill Formula C7H3BrF4O

 

ORDER

DOCUMENTATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty