Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

[2-(2-Methoxyethoxy)ethoxy]acetic acid(3,6,9-trioxadecanoic acid) for synthesis, Sigma-Aldrich®

Supplier: MilliporeSigma
Danger
Synonyms: [2-(2-Methoxyethoxy)ethoxy]acetic acid3,6,9-Trioxadecanoic acid2-(2-(2-Methoxyethoxy)ethoxy)acetic acid

M-Clarity™ quality level = MQ200

8.43910.0250 8.43910.0050 8.43910.0250 8.43910.0050
EM8.43910.0050CS 12735 USD InStock
EM8.43910.0050 CA8.43910.0050 CA8.43910.0250 EM8.43910.0250
[2-(2-Methoxyethoxy)ethoxy]acetic acid(3,6,9-trioxadecanoic acid) for synthesis, Sigma-Aldrich®
[2-(2-Methoxyethoxy)ethoxy]acetic acid (3,6,9-Trioxadecanoic acid)


Formula: C₇H₁₄O₅
MW: 178.1831 g/mol
MDL Number: MFCD00044098
CAS Number: 16024-58-1
EINECS: 240-162-1
ADR: 8,III
Order Now
Specification Test Results
Molar Mass 178.18 g/mol
Boiling Point 140 °C (2 mbar)
Flash Point 160 °C
Density 1.157 g/cm3
pH 1.8 (100 g/l, H2O)

 

ORDER

DOCUMENTATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty