Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Ammonium tetrafluoroborate for synthesis, Sigma-Aldrich®

Supplier: MilliporeSigma
Danger

M-Clarity™ quality level = MQ200

8.43945.1000 8.43945.0100 8.43945.9025 8.43945.9025 8.43945.0100 8.43945.1000
EM8.43945.9025EA 1753 USD InStock
CA8.43945.0100 EM8.43945.9025 EM8.43945.1000 EM8.43945.0100 CA8.43945.9025 CA8.43945.1000
Ammonium tetrafluoroborate for synthesis, Sigma-Aldrich®
Ammonium tetrafluoroborate


Formula: H₄BF₄N
MW: 104.84 g/mol
MDL Number: MFCD06245785
CAS Number: 13826-83-0
UN: 3260
ADR: 8,II
Order Now
Specification Test Results
Density 1.85 g/cm3 (20 °C)
pH 3.5 (5 g/l, H2O, 20 °C)

 

ORDER

DOCUMENTATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty