Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Scheduled maintenance of the site is planned from June 8th at 7:00 PM to June 9th at 5:00 PM ET. During this time, contract pricing and registration functionality will be unavailable. We appreciate your patience.

Dichloromethane ≥99.5%, GR ACS, Supelco®

Supplier: MilliporeSigma
Warning
Synonyms: Methane dichlorideMethylene chloride

DX0835-8 DX0835-8 DX0835-3 DX0835-6 DX0835-6 DX0835-5 DX0835-5 DX0835-3 DX0835P-4 DX0835-20 DX0835P-4 DX0835-20
CADX0835-3CS 5081.19 USD InStock
CADX0835-3 EM-DX0835-6 EM-DX0835-8 EM-DX0835-20 CADX0835-8 CADX0835-5 CADX0835-6 EM-DX0835-5 EMD-DX0835P-4 CADX0835-20 EMD-DX0835-3 CADX0835P-4
Dichloromethane ≥99.5%, GR ACS, Supelco®
Dichloromethane
Meets ACS Specifications, Meets Reagent Specifications for testing USP/NF monographs


Formula: CH₂Cl₂
MW: 84.93 g/mol
Boiling Pt: 39.8 °C (1013 hPa)
Melting Pt: –95 °C
Density: 1.322 g/cm³ (20 °C)
Storage Temperature: Ambient
MDL Number: MFCD00000881
CAS Number: 75-09-2
EINECS: 200-838-9
UN: 1593
ADR: 6.1,III
Merck Index: 12,06140
https://us-legacy.vwr.com/stibo/web/std.lang.all/14/30/20961430.jpg
Order Now
Specification Test Results
Assay (GC) 99.5% min
Appearance Clear
Color (APHA) 10 max
Free halogens To pass test
Infrared spectrum Conforms to standard
Residue after evaporation 0.002% max
Titrable acid 0.0003meq/g max
Water (H2O) 0.02% max

 

ORDER

DOCUMENTATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty