Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Hydrochloric acid 36.5 - 38.0%, GR ACS, Supelco®

Supplier: MilliporeSigma
Synonyms: Muriatic acid

M-Clarity™ quality level = MQ200

HX0603-4 HX0603-4 HX0603-75 HX0603P-5 HX0603-75 HX0603-3 HX0603-23 HX0603-23 HX0603P-5 HX0603-3
EM-HX0603P-5CS 618 USD InStock
CAHX0603-75 EM-HX0603P-5 CAHX0603-23 CAHX0603-3 CAHX0603P-5 EMD-HX0603-3 CAHX0603-4 EMD-HX0603-4 EM-HX0603-23 EM-HX0603-75
Hydrochloric acid 36.5 - 38.0%, GR ACS, Supelco®
Hydrochloric acid
Meets ACS Specifications, Meets Reagent Specifications for testing USP/NF monographs


Formula: HCl
MW: 36.46 g/mol
Boiling Pt: 110 °C (1013 hPa)
Melting Pt: –30 °C
Density: 1.18 g/cm³ (20 °C)
Storage Temperature: Ambient
MDL Number: MFCD00011324
CAS Number: 7647-01-0
EINECS: 231-595-7
UN: 1789
ADR: 8,II
Order Now
Specification Test Results
Aluminum (Al) 0.3ppm max
Ammonium (NH4) 3ppm max
Appearance To pass test
Arsenic (As) 0.01ppm max
Assay 36.5-38.0%
Bromide (Br) 0.005% max
Calcium (Ca) 0.3ppm max
Chromium (Cr) 0.2ppm max
Color (APHA) 10 max
Copper (Cu) 0.1ppm max
Free chlorine (as Cl) 1ppm max
Gold (Au) 0.3ppm max
Heavy metals (as Pb) 1ppm max
Iron (Fe) 0.2ppm max
Lead (Pb) 0.1ppm max
Magnesium (Mg) 0.3ppm max
Manganese (Mn) 0.3ppm max
Mercury (Hg) 0.005ppm max
Nickel (Ni) 0.1ppm max
Potassium (K) 0.3ppm max
Residue after ignition 5ppm max
Sodium (N/A) 0.3ppm max
Sulfate (SO4) 1ppm max
Sulfite (SO3) 1ppm max
Tin (Sn) 0.3ppm max
Titanium (Ti) 0.3ppm max
Zinc (Zn) 0.3ppm max

 

ORDER

DOCUMENTATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty