Would you like to log in again?
Low-Particle Bottles and Jugs, Thermo Scientific
Supplier: Thermo Scientific
114-250A/LP 113-500A/LP 114-125CT/LP 114-125A/LP 111-04A/LP 130-005/LP 110-80A/LP 114-060A/LP 114-250C/LP
EP114-125A-LPCS 408.72 USD InStock
EP114-125A-LP EP113-500ALP EP110-80A-LP EP114-250A-LP EP114-250C-LP 89026-624 89026-620 EP111-04A-LP 10031-388
Low-Particle Bottles and Jugs, Thermo Scientific
Bottles Glass Bottles
Glass containers are certified to contain fewer than 5 particles per ml larger than 0.5 µm.
  • Class 100/10 glass
  • Certificate of Analysis is included with each shipment
  • Available in clear or amber
  • Individual bottles double bagged to maintain integrity until point of use
  • 4 L jugs suitable for hazardous shipping in conjunction with combination packaging
  • Recommended for low-particle applications
Ideal for preparation and storage of standards, off-line D.I. water and chemical sampling for particle analysis, and storage and packaging of chemicals, reagents, and powders. Amber glass protects light-sensitive chemicals. Containers are assembled with low-shedding, polypropylene caps with chemically-inert fluoropolymer-resin faced liners that do not contain adhesives. Exceeds USP specifications for particle content of containers.
Ordering information: All containers are double-bagged in a ISO Class 5 (FED-STD-209E Class 100/M3.5) cleanroom. Supplied with Certificate of Analysis.
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty