Would you like to log in again?
Chemical Preservative Vials, Thermo Scientific
Supplier: Thermo Scientific
PP140-40CEP.5HA
EP14040CEP.5HACS 357.24 USD InStock
EP14040CEP.5HA
Chemical Preservative Vials, Thermo Scientific
Vials
Chemically preserved Environmental sample containers save the time, tedium and chemical contact associated with diluting and aliquoting chemical preservatives.
  • Simplify field preservation
  • Reduces the risk of container and preservative contamination
Certifications: Certified to meet the EPA Performance Based Specifications for indicated analyses; packaged in compliance with the shipping requirements of 49 CFR Part 173.4 DOT Small Quantities Classification; manufactured under an ISO certified quality management system.
Delivery information: Includes Certified sample containers prepreserved with chemical preservative; certificate of analysis for container and preservative, individually barcoded containers traceable to certs, sample analysis labels unattached for optional use, hazardous material compliant packaging. MSDS available upon request.
Order Now
ORDER
DOCUMENTATION
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty