Would you like to log in again?
Economy Certified Glass VOA Vials with 0.125" Septa, Thermo Scientific
Supplier: Thermo Scientific
GVB-100A GVB-100C
EPGVB100ACS 241.94 USD InStock
EPGVB100A EPGVB-100C
Economy Certified Glass VOA Vials with 0.125" Septa, Thermo Scientific
Vials
Septa vials with all the essentials and none of the frills.
  • Vial shoulder slope optimized to prevent air bubble entrapment
  • Septum permanently bonded to the cap for a secure seal
Clear or amber vials; Amber is recommended for light-sensitive analytes. Open-top white polypropylene cap with standard 0.125" thick PTFE-lined silicone septum.
Thermo Scientific Economy Certified VOA Vials with 0.125" Septa are designed for water sample collection for volatile organic analysis (VOA). You get our standard 40 ml VOA vials, processed and Certified to meet EPA Performance Based Specifications for Volatile Organic Analysis. No bar coding or other add-ons keeps costs low while eliminating the need to wash and certify vials in-house.
Ordering information: Certificate of analysis included in each case.
Order Now
ORDER
ACCESSORIES
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty