Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

EP Scientific Low-Carbon Total Organic Carbon Vials, Tubes and Bottles, Thermo Scientific

Supplier: Thermo Scientific

REVIEWS (0)

Total Ratings: 0
Avg. Ratings: out of 5

Sort all by:

40A-TOC/DB/LL 40C-TOC/DB S114-250C/TOC 40C-TOC/LL 40C-TOC/DB/LL SCT-18100/TOC S114-250A/TOC S114-125A/TOC 1000A/TOC CT18-TOC/LL 40C-TOC 3115-OTWS-2
EPSCT18100TOCCS 1419.63 USD InStock
EPSCT18100TOC 10142-542 EP1000A-TOC EPCT-18-TOC-LL EPS114-125ATOC EP40C-TOC EP40C-TOC-LL EP40C-TOC-DBLL EP40C-TOC-DB EPS114250CTOC EP40A-TOCDBLL EPS114-250ATOC
EP Scientific Low-Carbon Total Organic Carbon Vials, Tubes and Bottles, Thermo Scientific
Bottles Glass Bottles
Containers are certified to contain low levels of carbon and are for use in Total Organic Carbon (TOC) testing and sampling. Ideal for cleaning validation, off-line sampling and grab sampling of high purity water, and for preparation and storage of standards.

  • Available in several sizes and styles of containers, including the popular 40 ml autosampler vials
  • Cleaned, certified and ready to use

All containers have open-top caps, unless otherwise specified, and fluoropolymer resin-lined septa.

Manufactured, tested and packaged by exacting standards, Thermo Scientific™ TOC Certified Containers are certified to contribute <10 ppb or <20 ppb TOC as background.

Vials feature a septum designed to stay inside the cap while sampling and are appropriate for use as autosampler vials or for sampling.

Ordering information: Supplied with Certificate of Analysis.

Order Now
 

ORDER

ACCESSORIES

DOCUMENTATION

WRITE A REVIEW

REVIEWS (0)

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty