Would you like to log in again?
EP Scientific Silanized Culture Tubes and Vials, Thermo Scientific
Supplier: Thermo Scientific
REVIEWS (0)
Total Ratings: 0
Avg. Ratings: out of 5
Sort all by:
STT-16100-S STT-13100-S SCA-SV2-2 CTS-1275 CTS-13100 CTS-16100 CTS-16125-2 STT-16125-S
EPCTS-16125-2CS 779.39 USD InStock
EPCTS-16125-2 EPSTT-16100-S EPSCA-SV2-2 EPCTS-13100 EPCTS-16100 EPSTT-16125-S EPSTT-13100-S EPCTS-1275
EP Scientific Silanized Culture Tubes and Vials, Thermo Scientific
Tubes Culture Tubes
Ready-to-use, disposable glassware is ideal for trace organic analysis and storage of materials prone to adhering to glass.
Tubes EPSTT-13100-S and EPSTT-16100-S have threads for caps (not supplied).
Choose from Type 1 borosilicate glass culture tubes or autosampler vials with open-top caps and septa. Certificate of Conformance included.
All tubes and vials, except EPCTS-13100-D, feature a methylsilylating agent introduced by vapor phase deposition onto the surface of the glassware. This treatment inhibits materials from adhering to the surface of the container, allowing for maximum recovery of trace analyte. Tubes EPCTS-13100-D exclude the vapor phase deposition process and use instead a dip process.
Ordering information: All tubes and vials are clear glass. For amber glass, contact your Avantor sales representative.
Order Now
ORDER
WRITE A REVIEW
REVIEWS (0)
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty