Would you like to log in again?
Ultragrade 19 Vacuum Pump Oil, Edwards
Supplier: Edwards
H11025015 H11025015
EVH110-25-015EA 64 USD InStock
CAH110-25-015 EVH110-25-015
Ultragrade 19 Vacuum Pump Oil, Edwards
Lubricants
This oil has been produced by a unique hydrotreating process
When combined with additives, the Ultragrade® oils give improved vacuum performance, thermal and oxidation stability and rust inhibition. The anti-oxidants included in the oils enable the pump to withstand high operating temperatures without oil degradation, so extending intervals between oil changes.
This process eliminates nitrogen, sulphur and oxygen and the aromatic hydrocarbons, leaving a clear base stock essentially free of impurities.
Pour Point: -16°
Order Now
ORDER
SPECIFICATIONS
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty