Would you like to log in again?
Human Tyrosinase (206-214) Acetate Salt
Supplier: Bachem
4040740.0001 4040740.0005
H-3848.0005BAEA 609.62 USD InStock
H-3848.0005BA H-3848.0001BA
Human Tyrosinase (206-214) Acetate Salt
Enzymes
Sequence: H-Ala-Phe-Leu-Pro-Trp-His-Arg-Leu-Phe-OH
AFLPWHRLF is a tyrosinase epitope recognized by HLA-A24 restricted, tumor-infiltrating lymphocytes (TIL). Therefore it represents a reagent that may be used to generate melanoma-specific T cells for adoptive immunotherapy, as well as in peptide vaccines for HLA-A24+ melanoma patients.
Order Now
ORDER
SPECIFICATIONS
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty