Would you like to log in again?
H-Met-Arg-Phe-Ala-OH
Supplier: BACHEM AMERICAS INC
REVIEWS (0)
Total Ratings: 0
Avg. Ratings: out of 5
Sort all by:
4004330.0050 4004330.0250
H-4320.0050BAEA 433.14 USD InStock
H-4320.0050BA H-4320.0250BA
H-Met-Arg-Phe-Ala-OH
Proteins and Peptides
MRFA is used as a calibration standard in mass spectrometry (ESI). Miao et al. studied Pt(II) complexes of the tetrapeptide by mass spectrometric methods. MRFA has been shown to be a competitive inhibitor of an enkephalin-generating endopeptidase isolated from rat brain. The peptide is a substrate for dipeptidyl peptidase III from human erythrocytes and for snapalysin.
Order Now
ORDER
SPECIFICATIONS
WRITE A REVIEW
REVIEWS (0)
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty