Would you like to log in again?
Acetyl-(D-Arg¹⁰,Cys¹¹,D-Phe¹⁴,Cys¹⁷)-beta-MSH (10-17) Amide Trifluoroacetate
Supplier: Bachem
4049312.0005 4049312.0001
H-6268.0005BAEA 809.49 USD InStock
H-6268.0005BA H-6268.0001BA
Acetyl-(D-Arg¹⁰,Cys¹¹,D-Phe¹⁴,Cys¹⁷)-beta-MSH (10-17) Amide Trifluoroacetate
Proteins and Peptides
Highly potent and selective β-MSH-derived melanocortin-4 receptor (MC-4R) agonist with sub-nanomolar potency. The binding affinities for human MC-4R, MC-1R, MC-3R and MC-5R were 0.55 ± 0.06, 16.78 ± 1.53, 56.79 ± 4.66, and < 500 nM, respectively. Daily subcutaneous injection of H-6268 to diet-induced obese rats over a period of 14 days demonstrated an effect on reduction of fat mass.
Order Now
ORDER
SPECIFICATIONS
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty