Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Chloramphenicol

Supplier: IBI Scientific
Synonyms: Chloromycetin®(R,R)-ChloramphenicolTega-Cetin

IB02080
IB02080EA 53.23 USD InStock
IB02080
Chloramphenicol
Chloramphenicol
Chloramphenicol is used in the amplification of plasmids and colony hybridizations

Refrigerated shipping and storage is required for this product.

Chlroamphenicol is used at the following concentrations: Stringent plasmids (25µg/ml) and Relaxed Plasmids (170µg/ml). Antibiotic inhibition/sensitivity assay, Crystallinity and Chromatographic Purity: PASS.


Formula: Cl₂CHCONHCH(CH₂OH)CH(OH)C₆H₄NO₂
MW: 323.13 g/mol
MDL Number: MFCD00078159
CAS Number: 56-75-7
Order Now

 

ORDER

DOCUMENTATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty