Would you like to log in again?
QS710 Horizontal Gel Electrophoresis Unit, IBI Scientific
Supplier: IBI Scientific
IB51000
IB51000EA 599.79 USD InStock
IB51000
QS710 Horizontal Gel Electrophoresis Unit, IBI Scientific
Electrophoresis Systems Horizontal Gel Electrophoresis Systems
Designed for quick screening and accurate, rapid separation of nucleic acids
Well visualization stripes improve visibility, facilitating gel loading. Vented lid reduces condensation, improving resolution of fast runs. Gel trays transmit 312nm UV light while allowing maximum sample capacity using multiple combs, or maximum running lengths using a single comb.
Electrical gravity connector minimizes electrical hazards and eliminates sample disturbance during setup. Safety-tipped power cords with retractable sleeves and plastic tips reduce electrical hazards.
Precision-made Delrin® analytical combs (sold separately) are available in a wide range of thicknesses and tooth configurations to accommodate multiple samples or large sample volumes.
Use for electrophoresis procedures that do not require buffer recirculation. Unit features corner leveling feet for perfectly straight gel runs, three-piece acrylic construction that resists warping, and a leveling bubble to ensure perfect gels. Three gel sizes allow applications ranging from quick screens to high-resolution separations.
Ordering information: System includes a buffer chamber; casting system with tray; leveling bubble; safety cover and one pair of power cords; two 1.5mm thick, 8-tooth combs; and operator’s manual.
Order Now
ORDER
SPECIFICATIONS
ACCESSORIES
DOCUMENTATION
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty