Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Bromophenol blue sodium salt ≥98% for molecular biology

Supplier: IBI Scientific
Synonyms: Sodium 2,6-dibromo-4-(3-(3,5-dibromo-4-hydroxyphenyl)-1,1-dioxido-3H-benzo[c][1,2]oxathiol-3-yl)phenolate4,4'-(3H-2,1-Benzoxathiol-3-ylidene)bis[2,6-dibromophenol] S,S-dioxide sodium saltBromophenol blue water solubleSodium α-(3,5-dibromo-4-oxo-2,5-cyclohexadienylidene)-α-(3,5-dibromo-4-hydroxyphenyl)toluenesulphonateSodium α-(3,5-dibromo-4-oxo-2,5-cyclohexadienylidene)-α-(3,5-dibromo-4-hydroxyphenyl)toluenesulfonate

IB74040
IB74040EA 76.58 USD InStock
IB74040
Bromophenol blue sodium salt ≥98% for molecular biology
Bromophenol blue sodium salt
Bromophenol Blue is used as a tracking dye to monitor the progress of electrophoresis separations. Bromophenol Blue usually comigrates with DNA molecules around 0.5kb. Therefore, bromophenol blue provides an index of the mobility of the fastest fragments and is valuable in determining the length of the gel, over which the separation of DNA has occurred.

Store at room temperature, keep tightly sealed, and protect from moisture and light.


Formula: C₁₉H₉Br₄NaO₅S
MW: 691.95 g/mol
MDL Number: MFCD00013793
CAS Number: 34725-61-6
Order Now
Specification Test Results
Purity 98.0%
pH (1.0M water) 6.6-7.4
Insolubles 0.01%

 

ORDER

DOCUMENTATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty