Would you like to log in again?
KODAK® X-OMAT® LS-1 (XLS) Autoradiography Film, Carestream Health
Supplier: CARESTREAM HEALTH
KODAK® X-OMAT®
864-6770 161-1342
IB161-1342CS 1134.75 USD InStock
IB161-1342 IB8646770
KODAK® X-OMAT® LS-1 (XLS) Autoradiography Film, Carestream Health
Autoradiography Films Films
An economical, blue-tinted film for use with commonly used isotopes and chemiluminescence
Provides significantly lower sensitivity and longer exposure times compared to BioMax® MR, BioMax® MS, and XAR films. A double-coated emulsion film suitable for manual and automated processing. Use with 7.5 or 15W bulb.
Ordering information: Non-interleaved packaging.
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty