Synonyms:
Acetic acid ammonium salt
0219384891
0219384880
0219384890
IC0219384891EA
174.07
USD
InStock
IC0219384891
IC0219384880
IC0219384890
Ammonium acetate ≥97%, Ultrapure
Ammonium acetate
Ammonium acetate is a chemical compound which is derived from the reaction of ammonia and acetic acid. Ammonium acetate is volatile at low pressures. Because of which it has been used to replace cell buffers with non-volatile salts, in preparing samples for mass spectrometry. It is used as a buffer for mobile phases for HPLC with ELSD detection.
Ammonium acetate Ultra pure is for applications which require tight control of elemental content. Ammonium acetate is a widely used reagent in molecular biology and chromatography. Its applications include the purification and precipitation of DNA and protein crystallization. Ammonium acetate is commonly used in HPLC and MS analysis of various compounds, such as oligosaccharides, proteins, and peptides. Ammonium acetate is also used for the nonaqueous capillary electrophoresis-mass spectrometry (NACE-MS) of lipophilic peptides and therapeutic drugs.